missing translation for 'onlineSavingsMsg'
Learn More

2',4',6'-Trimethylacetophenone 97.0+%, TCI America™

Catalog No. p-7136618
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11175ML 5 mL
T111725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T11175ML Supplier TCI America Supplier No. T11175ML
Only null left

Chemical Identifiers

CAS 1667-01-2
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00008735
InChI Key XWCIICLTKWRWCI-UHFFFAOYSA-N
Synonym 1-mesitylethanone, 2',4',6'-trimethylacetophenone, acetylmesitylene, acetomesitylene, mesityl methyl ketone, 1-2,4,6-trimethylphenyl ethanone, ethanone, 1-2,4,6-trimethylphenyl, 2-acetylmesitylene, 2,4,6-trimethylacetophenone, methyl 2,4,6-trimethylphenyl ketone
PubChem CID 15461
IUPAC Name 1-(2,4,6-trimethylphenyl)ethan-1-one
SMILES CC(=O)C1=C(C)C=C(C)C=C1C

Specifications

Color Yellow
Boiling Point 236°C
Quantity 5 mL
Formula Weight 162.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′,4′,6′-Trimethylacetophenone