missing translation for 'onlineSavingsMsg'
Learn More

2',4',6'-Trimethylacetophenone, 97+%

Catalog No. AAA1154914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1154914 25 g
AAA1154922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1154914 Supplier Thermo Scientific Chemicals Supplier No. A1154914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1667-01-2 | C11H14O | 162.23 g/mol

2',4',6'-Trimethylacetophenone is used as laboratory chemicals, manufacture of substances and intermediated for chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,4′,6′-Trimethylacetophenone is used as laboratory chemicals, manufacture of substances and intermediated for chemical synthesis.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 1667-01-2
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00008735
InChI Key XWCIICLTKWRWCI-UHFFFAOYSA-N
Synonym 1-mesitylethanone, 2',4',6'-trimethylacetophenone, acetylmesitylene, acetomesitylene, mesityl methyl ketone, 1-2,4,6-trimethylphenyl ethanone, ethanone, 1-2,4,6-trimethylphenyl, 2-acetylmesitylene, 2,4,6-trimethylacetophenone, methyl 2,4,6-trimethylphenyl ketone
PubChem CID 15461
IUPAC Name 1-(2,4,6-trimethylphenyl)ethanone
SMILES CC(=O)C1=C(C)C=C(C)C=C1C

Specifications

Density 0.975
Boiling Point 235°C to 236°C
Flash Point >110°C (230°F)
Refractive Index 1.517
Quantity 25 g
Beilstein 2044595
Solubility Information Difficult to mix.
Formula Weight 162.23
Percent Purity 98%
Chemical Name or Material 2',4',6'-Trimethylacetophenone
EINECSNumber 216-783-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only