missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethyl-1,6-heptadien-4-ol, 98%

Catalog No. p-7024849
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2008603 1 g
AAB2008606 5 g
AAB2008614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2008603 Supplier Thermo Scientific Chemicals Supplier No. B2008603

CAS: 79604-66-3 | C10H18O | 154.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79604-66-3
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00026060
InChI Key METKJWMTLIYYIQ-UHFFFAOYSA-N
Synonym 2,4,6-trimethyl-1,6-heptadien-4-ol, diisobutenyl methyl carbinol
PubChem CID 144898
IUPAC Name 2,4,6-trimethylhepta-1,6-dien-4-ol
SMILES CC(=C)CC(C)(CC(=C)C)O

Specifications

Quantity 1 g
Formula Weight 154.25
Percent Purity 98%
Chemical Name or Material 2,4,6-Trimethyl-1,6-heptadien-4-ol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only