missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',4',6'-Trimethoxyacetophenone, 98%
Description
2',4',6'-Trimethoxyacetophenone is used in the production of 4,2',4',6'-tetramethoxy-chalcone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2′,4′,6′-Trimethoxyacetophenone is used in the production of 4,2′,4′,6′-tetramethoxy-chalcone.
Solubility
Soluble in chloroform.
Notes
Incompatible with strong oxidizing agents.
2′,4′,6′-Trimethoxyacetophenone is used in the production of 4,2′,4′,6′-tetramethoxy-chalcone.
Solubility
Soluble in chloroform.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 101°C to 104°C |
| Quantity | 5 g |
| Beilstein | 519324 |
| Solubility Information | Soluble in chloroform. |
| Formula Weight | 210.23 |
| Percent Purity | 98% |
| Chemical Name or Material | 2',4',6'-Trimethoxyacetophenone |
Safety and Handling
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only