missing translation for 'onlineSavingsMsg'
Learn More

2',4',6'-Trimethoxyacetophenone, 98%

Catalog No. AAA1432806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1432806 5 g
AAA1432814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1432806 Supplier Thermo Scientific Chemicals Supplier No. A1432806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 832-58-6 | C11H14O4 | 210.23 g/mol

2',4',6'-Trimethoxyacetophenone is used in the production of 4,2',4',6'-tetramethoxy-chalcone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,4′,6′-Trimethoxyacetophenone is used in the production of 4,2′,4′,6′-tetramethoxy-chalcone.

Solubility
Soluble in chloroform.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 832-58-6
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.23
MDL Number MFCD00017238
InChI Key KPZWHZSIXZXDMW-UHFFFAOYSA-N
Synonym 2',4',6'-trimethoxyacetophenone, 1-2,4,6-trimethoxyphenyl ethanone, 2,4,6-trimethoxyacetophenone, ethanone, 1-2,4,6-trimethoxyphenyl, ethanone,1-2,4,6-trimethoxyphenyl, 1-acetyl-2,4,6-trimethoxybenzene, o-methylxanthoxylin, cambridge id 5135317, 2,4,6-trimethoxy acetophenone, 2/',4/',6/'-trimethoxyacetophenone
PubChem CID 123089
IUPAC Name 1-(2,4,6-trimethoxyphenyl)ethanone
SMILES COC1=CC(OC)=C(C(C)=O)C(OC)=C1

Specifications

Melting Point 101°C to 104°C
Quantity 5 g
Beilstein 519324
Solubility Information Soluble in chloroform.
Formula Weight 210.23
Percent Purity 98%
Chemical Name or Material 2',4',6'-Trimethoxyacetophenone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only