missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triisopropylbenzoic acid, 97%

CAS: 49623-71-4 | C16H24O2 | 248.366 g/mol

Supplier:  Thermo Scientific Chemicals A1548918

Quantity 50 g

 View more versions of this product

Catalog No. AAA1548918


Only null left

Description

Description

2,4,6-Triisopropylbenzoic acid is a hindered acid intermediate compound in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Triisopropylbenzoic acid is a hindered acid intermediate compound in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

49623-71-4
248.366
ULVHAZFBJJXIDO-UHFFFAOYSA-N
96214
CC(C)C1=CC(=C(C(=C1)C(C)C)C(=O)O)C(C)C
C16H24O2
MFCD00015031
2,4,6-triisopropylbenzoic acid, 2,4,6-triisopropyl benzoic acid, 2,4,6-tris 1-methylethyl benzoic acid, 2,4,6-tri propan-2-yl benzoic acid, benzoic acid,2,4,6-tris 1-methylethyl, benzoic acid, 2,4,6-tris 1-methylethyl, 2,4,6-tris methylethyl benzoic acid, acmc-20aoew, pubchem7877
2,4,6-tri(propan-2-yl)benzoic acid

Specifications

2,4,6-Triisopropylbenzoic acid
184°C to 187°C
50 g
2372331
Slightly soluble in water.
CC(C)C1=CC(=C(C(=C1)C(C)C)C(=O)O)C(C)C
248.366
248.37
49623-71-4
C16H24O2
MFCD00015031
2,4,6-triisopropylbenzoic acid, 2,4,6-triisopropyl benzoic acid, 2,4,6-tris 1-methylethyl benzoic acid, 2,4,6-tri propan-2-yl benzoic acid, benzoic acid,2,4,6-tris 1-methylethyl, benzoic acid, 2,4,6-tris 1-methylethyl, 2,4,6-tris methylethyl benzoic acid, acmc-20aoew, pubchem7877
ULVHAZFBJJXIDO-UHFFFAOYSA-N
2,4,6-tri(propan-2-yl)benzoic acid
96214
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 256-400-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions