missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trihydroxybenzoic acid monohydrate, 90+%

Catalog No. p-7026655
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2276814 25 g
AAB2276822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2276814 Supplier Thermo Scientific Chemicals Supplier No. B2276814

CAS: 71989-93-0 | C7H8O6 | 188.135 g/mol

2,4,6-Trihydroxybenzoic acid monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trihydroxybenzoic acid monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 71989-93-0
Molecular Formula C7H8O6
Molecular Weight (g/mol) 188.135
MDL Number MFCD00149095
InChI Key HWZIRFCGHAROOI-UHFFFAOYSA-N
Synonym 2,4,6-trihydroxybenzoic acid hydrate, 2,4,6-trihydroxybenzoic acid monohydrate, benzoic acid, 2,4,6-trihydroxy-, monohydrate, phloroglucinolcarboxylic acid, pubchem20337, acmc-209olb, 2,4,6-trihydroxybenzoicacidmonohydrate, 2,4,6-trihydroxybenzoic acid, hydrate, 2,4,6-trihydroxybenzoic acid monohydrate, technical t, 2,4,6-trihydroxybenzoic acid monohydrate, technical grade, predominantly 1,3,5-benzenetriol
PubChem CID 2723793
IUPAC Name 2,4,6-trihydroxybenzoic acid;hydrate
SMILES C1=C(C=C(C(=C1O)C(=O)O)O)O.O

Specifications

Melting Point ∼210°C (decomposition)
Quantity 25 g
Beilstein 2212148
Solubility Information Soluble in water.
Formula Weight 188.14 (170.12 Anhydrous)
Percent Purity 94%
Chemical Name or Material 2,4,6-Trihydroxybenzoic acid monohydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-467-5
RTECSNumber DH8910000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only