missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trihydroxybenzaldehyde, 95%

Catalog No. p-7024644
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1967803 1 g
AAA1967806 5 g
AAA1967814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1967803 Supplier Thermo Scientific Chemicals Supplier No. A1967803

CAS: 487-70-7 | C7H6O4 | 154.12 g/mol

2,4,6-Trihydroxybenzaldehyde is used in the preparation of 2-methyl-phloroglucinol by hydrogenation using 5% palladium/carbon. It acts as a reactant for the synthesis of biologically active molecules such as 2,4,6-trichlorophenyl hydrazones. It is also used as a xanthine oxidase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trihydroxybenzaldehyde is used in the preparation of 2-methyl-phloroglucinol by hydrogenation using 5% palladium/carbon. It acts as a reactant for the synthesis of biologically active molecules such as 2,4,6-trichlorophenyl hydrazones. It is also used as a xanthine oxidase inhibitor.

Solubility
Soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and strong bases.

Chemical Identifiers

CAS 487-70-7
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00003329
InChI Key BTQAJGSMXCDDAJ-UHFFFAOYSA-N
Synonym phloroglucinaldehyde, benzaldehyde, 2,4,6-trihydroxy, formylphloroglucinal, phloroglucinolcarboxaldehyde, formylphloroglucinol, phloroglucinol aldehyde, 2,4,6-trihydroxy-benzaldehyde, 2,4,6-trihydroxybenzaldehyd, pubchem21790, acmc-1ao01
PubChem CID 68099
IUPAC Name 2,4,6-trihydroxybenzaldehyde
SMILES OC1=CC(O)=C(C=O)C(O)=C1

Specifications

Melting Point ∼195°C (decomposition)
Quantity 1 g
Beilstein 2254429
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 154.12
Percent Purity 95%
Chemical Name or Material 2,4,6-Trihydroxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-663-7
RTECSNumber CU8440000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only