missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluoropyrimidine, 98%

CAS: 696-82-2 | C4HF3N2 | 134.06 g/mol

Supplier:  Thermo Scientific Chemicals L0440406

 View more versions of this product

Catalog No. AAL0440406

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4,6-Trifluoropyrimidine
100°C to 102°C
1.405
5 g
607190
pyrimidine, 2,4,6-trifluoro, pubchem6891, 2,4,6-trifluoropyrimide, 2,4, 6-trifluoropyrimidine, pyrimidine,2,4,6-trifluoro, pyrimidine, 2,4,6-trifluoro-6ci,7ci,8ci,9ci
FC1=CC(F)=NC(F)=N1
134.06
134.06
696-82-2
C4HF3N2
MFCD00039705
UN3265
Moisture sensitive
NTSYSQNAPGMSIH-UHFFFAOYSA-N
2,4,6-trifluoropyrimidine
69677
98%

Chemical Identifiers

696-82-2
134.06
NTSYSQNAPGMSIH-UHFFFAOYSA-N
69677
FC1=CC(F)=NC(F)=N1
C4HF3N2
MFCD00039705
pyrimidine, 2,4,6-trifluoro, pubchem6891, 2,4,6-trifluoropyrimide, 2,4, 6-trifluoropyrimidine, pyrimidine,2,4,6-trifluoro, pyrimidine, 2,4,6-trifluoro-6ci,7ci,8ci,9ci
2,4,6-trifluoropyrimidine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 211-801-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only