missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluoropyrimidine, 98%

Catalog No. AAL0440403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0440403 1 g
AAL0440406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0440403 Supplier Thermo Scientific Chemicals Supplier No. L0440403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 696-82-2 | C4HF3N2 | 134.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 696-82-2
Molecular Formula C4HF3N2
Molecular Weight (g/mol) 134.06
MDL Number MFCD00039705
InChI Key NTSYSQNAPGMSIH-UHFFFAOYSA-N
Synonym pyrimidine, 2,4,6-trifluoro, pubchem6891, 2,4,6-trifluoropyrimide, 2,4, 6-trifluoropyrimidine, pyrimidine,2,4,6-trifluoro, pyrimidine, 2,4,6-trifluoro-6ci,7ci,8ci,9ci
PubChem CID 69677
IUPAC Name 2,4,6-trifluoropyrimidine
SMILES FC1=CC(F)=NC(F)=N1

Specifications

Boiling Point 100°C to 102°C
Refractive Index 1.405
Quantity 1 g
UN Number UN3265
Beilstein 607190
Sensitivity Moisture sensitive
Formula Weight 134.06
Percent Purity 98%
Chemical Name or Material 2,4,6-Trifluoropyrimidine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 211-801-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only