missing translation for 'onlineSavingsMsg'
Learn More

2',4',6'-Trifluoroacetophenone 98.0+%, TCI America™

Catalog No. p-7137381
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27335G 5 g
T273325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27335G Supplier TCI America Supplier No. T27335G
Only null left

Chemical Identifiers

CAS 51788-77-3
Molecular Formula C8H5F3O
Molecular Weight (g/mol) 174.12
MDL Number MFCD00061194
InChI Key FFJSZHCBWFPSJI-UHFFFAOYSA-N
Synonym 2',4',6'-trifluoroacetophenone, 1-2,4,6-trifluorophenyl ethanone, 2,4,6-trifluoroacetophenone, 1-2,4,6-trifluorophenyl ethan-1-one, ethanone, 1-2,4,6-trifluorophenyl, 1-2,4,6-trifluoro-phenyl-ethanone, 1-acetyl-2,4,6-trifluorobenzene, pubchem4290, acmc-209kvx, 2',4,6'-trifluoroacetophenone
PubChem CID 2776906
IUPAC Name 1-(2,4,6-trifluorophenyl)ethan-1-one
SMILES CC(=O)C1=C(F)C=C(F)C=C1F

Specifications

Color Yellow
Boiling Point 190°C
Quantity 5 g
Formula Weight 174.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′,4′,6′-Trifluoroacetophenone