missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichloropyrimidine 98.0+%, TCI America™

Catalog No. T041725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T041725G 25 g
T0417500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T041725G Supplier TCI America Supplier No. T041725G

Chemical Identifiers

CAS 3764-01-0
Molecular Formula C4HCl3N2
Molecular Weight (g/mol) 183.416
MDL Number MFCD00006063
InChI Key DPVIABCMTHHTGB-UHFFFAOYSA-N
Synonym pyrimidine, 2,4,6-trichloro, 2,4,6-trichloro pyrimidine, 2,4,6-trichloro-pyrimidine, ccris 7597, 2,4,6-trichlorpyrimidin, pubchem9310, acmc-1agpv, 2,6-trichloropyrimidine, 2.4.6-trichloropyrimidin, dsstox_cid_29143
PubChem CID 77378
IUPAC Name 2,4,6-trichloropyrimidine
SMILES C1=C(N=C(N=C1Cl)Cl)Cl

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 183.42
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 2,4,6-Trichloropyrimidine
EINECSNumber (5)-0929
TSCA Yes