missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichloropyrimidine, 98%

Catalog No. p-7020775
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1284206 5 g
AAA1284214 25 g
AAA1284222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1284206 Supplier Thermo Scientific Chemicals Supplier No. A1284206
Only null left

CAS: 3764-01-0 | C4HCl3N2 | 183.416 g/mol

It is applied In selective Suzuki coupling with arylboronic acids has been reported to give 6-arylpyrimidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied In selective Suzuki coupling with arylboronic acids has been reported to give 6-arylpyrimidines.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 3764-01-0
Molecular Formula C4HCl3N2
Molecular Weight (g/mol) 183.416
MDL Number MFCD00006063
InChI Key DPVIABCMTHHTGB-UHFFFAOYSA-N
Synonym pyrimidine, 2,4,6-trichloro, 2,4,6-trichloro pyrimidine, 2,4,6-trichloro-pyrimidine, ccris 7597, 2,4,6-trichlorpyrimidin, pubchem9310, acmc-1agpv, 2,6-trichloropyrimidine, 2.4.6-trichloropyrimidin, dsstox_cid_29143
PubChem CID 77378
IUPAC Name 2,4,6-trichloropyrimidine
SMILES C1=C(N=C(N=C1Cl)Cl)Cl

Specifications

Melting Point 21°C to 25°C
Density 1.595
Boiling Point 213°C to 215°C
Flash Point >110°C (230°F)
Odor Irritating
Refractive Index 1.57
Quantity 5 g
UN Number UN1759
Beilstein 118284
Solubility Information Insoluble in water.
Formula Weight 183.43
Percent Purity 98%
Chemical Name or Material 2,4,6-Trichloropyrimidine
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 223-183-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only