Learn More
2,4,6-Trichloropyridine, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 16063-69-7 |
|---|---|
| Molecular Formula | C5H2Cl3N |
| Molecular Weight (g/mol) | 182.44 |
| MDL Number | MFCD00971928 |
| InChI Key | FJNNGKMAGDPVIU-UHFFFAOYSA-N |
| Synonym | pyridine, 2,4,6-trichloro, 2,4,6-trichloro-pyridine, pubchem11020, acmc-1bu24, ksc497g8t, 2,4,6-tris chloranyl pyridine, 2,4,6-trichloropyridine |
| PubChem CID | 27665 |
| IUPAC Name | 2,4,6-trichloropyridine |
| SMILES | C1=C(C=C(N=C1Cl)Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC399590010
|
Thermo Scientific Chemicals
399590010 |
1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC399590050
|
Thermo Scientific Chemicals
399590050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 16063-69-7 | |
| 182.44 | |
| FJNNGKMAGDPVIU-UHFFFAOYSA-N | |
| 27665 | |
| C1=C(C=C(N=C1Cl)Cl)Cl |
| C5H2Cl3N | |
| MFCD00971928 | |
| pyridine, 2,4,6-trichloro, 2,4,6-trichloro-pyridine, pubchem11020, acmc-1bu24, ksc497g8t, 2,4,6-tris chloranyl pyridine, 2,4,6-trichloropyridine | |
| 2,4,6-trichloropyridine |
Specifications
| 16063-69-7 | |
| 100.0 | |
| Authentic | |
| Glass bottle | |
| MFCD00971928 | |
| pyridine, 2,4,6-trichloro, 2,4,6-trichloro-pyridine, pubchem11020, acmc-1bu24, ksc497g8t, 2,4,6-tris chloranyl pyridine, 2,4,6-trichloropyridine | |
| C1=C(C=C(N=C1Cl)Cl)Cl | |
| 182.44 | |
| 182.44 | |
| 2, 4, 6-Trichloropyridine |
| 97.5 | |
| >110°C | |
| 97.5% min. (GC) | |
| C5H2Cl3N | |
| 1g | |
| FJNNGKMAGDPVIU-UHFFFAOYSA-N | |
| 2,4,6-trichloropyridine | |
| 27665 | |
| 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
RUO â Research Use Only