missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichloropyridine, 98%

Catalog No. AC399590050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC399590050 5 g Glass bottle
AC399590010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399590050 Supplier Thermo Scientific Chemicals Supplier No. 399590050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16063-69-7 | C5H2Cl3N | 182.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16063-69-7
Molecular Formula C5H2Cl3N
Molecular Weight (g/mol) 182.44
MDL Number MFCD00971928
InChI Key FJNNGKMAGDPVIU-UHFFFAOYSA-N
Synonym pyridine, 2,4,6-trichloro, 2,4,6-trichloro-pyridine, pubchem11020, acmc-1bu24, ksc497g8t, 2,4,6-tris chloranyl pyridine, 2,4,6-trichloropyridine
PubChem CID 27665
IUPAC Name 2,4,6-trichloropyridine
SMILES C1=C(C=C(N=C1Cl)Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 33°C to 37°C
Color Brown-Yellow to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 182.44
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4, 6-Trichloropyridine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only