Learn More
2,4,6-Trichloropyridine, 97%
CAS: 16063-69-7 | C5H2Cl3N | 182.428 g/mol
$192.42 - $912.66
Chemical Identifiers
| CAS | 16063-69-7 |
|---|---|
| Molecular Formula | C5H2Cl3N |
| Molecular Weight (g/mol) | 182.428 |
| MDL Number | MFCD00971928 |
| InChI Key | FJNNGKMAGDPVIU-UHFFFAOYSA-N |
| Synonym | pyridine, 2,4,6-trichloro, 2,4,6-trichloro-pyridine, pubchem11020, acmc-1bu24, ksc497g8t, 2,4,6-tris chloranyl pyridine, 2,4,6-trichloropyridine |
| PubChem CID | 27665 |
| IUPAC Name | 2,4,6-trichloropyridine |
| SMILES | C1=C(C=C(N=C1Cl)Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5678706
|
Thermo Scientific Chemicals
H5678706 |
5 g |
Each for $192.42
|
|
|||||
|
AAH5678714
|
Thermo Scientific Chemicals
H5678714 |
25 g |
Each for $912.66
|
|
|||||
Description
It is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as pharmaceutical intermediates.
Solubility
Sparingly soluble in water. (0.113 mm Hg at 25°C)
Notes
Store at 2-8°C. Avoid using oxidizing agents.
Chemical Identifiers
| 16063-69-7 | |
| 182.428 | |
| FJNNGKMAGDPVIU-UHFFFAOYSA-N | |
| 27665 | |
| C1=C(C=C(N=C1Cl)Cl)Cl |
| C5H2Cl3N | |
| MFCD00971928 | |
| pyridine, 2,4,6-trichloro, 2,4,6-trichloro-pyridine, pubchem11020, acmc-1bu24, ksc497g8t, 2,4,6-tris chloranyl pyridine, 2,4,6-trichloropyridine | |
| 2,4,6-trichloropyridine |
Specifications
| 16063-69-7 | |
| >110°C (230°F) | |
| MFCD00971928 | |
| pyridine, 2,4,6-trichloro, 2,4,6-trichloro-pyridine, pubchem11020, acmc-1bu24, ksc497g8t, 2,4,6-tris chloranyl pyridine, 2,4,6-trichloropyridine | |
| FJNNGKMAGDPVIU-UHFFFAOYSA-N | |
| 2,4,6-trichloropyridine | |
| 27665 | |
| 97% |
| 33°C to 37°C | |
| C5H2Cl3N | |
| 5 g | |
| Sparingly soluble in water. (0.113 mm Hg at 25°C) | |
| C1=C(C=C(N=C1Cl)Cl)Cl | |
| 182.428 | |
| 182.44 | |
| 2,4,6-Trichloropyridine |
Safety and Handling
GHS H Statement
H301-H318-H317
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.
P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P363-P501c
H302-H317-H318
RTECSNumber : UU0875000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only