missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trichloropyridine, 97%
Description
It is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediates.
Solubility
Sparingly soluble in water. (0.113 mm Hg at 25°C)
Notes
Store at 2-8°C. Avoid using oxidizing agents.
It is used as pharmaceutical intermediates.
Solubility
Sparingly soluble in water. (0.113 mm Hg at 25°C)
Notes
Store at 2-8°C. Avoid using oxidizing agents.
Specifications
Specifications
| Melting Point | 33°C to 37°C |
| Flash Point | >110°C (230°F) |
| Quantity | 5 g |
| Solubility Information | Sparingly soluble in water. (0.113 mm Hg at 25°C) |
| Formula Weight | 182.44 |
| Percent Purity | 97% |
| Chemical Name or Material | 2,4,6-Trichloropyridine |
Safety and Handling
| Hazard Category | H302-H317-H318 |
| Hazard Statement | GHS H Statement H301-H318-H317 Toxic if swallowed. Causes serious eye damage. May cause an allergic skin reaction. |
| Precautionary Statement | P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P363-P501c |
| RTECSNumber | UU0875000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only