missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichloroaniline 98.0+%, TCI America™

Catalog No. T037525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T037525G 25 g
T0375500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T037525G Supplier TCI America Supplier No. T037525G

Chemical Identifiers

CAS 634-93-5
Molecular Formula C6H4Cl3N
Molecular Weight (g/mol) 196.455
MDL Number MFCD00007663
InChI Key NATVSFWWYVJTAZ-UHFFFAOYSA-N
Synonym s-trichloroaniline, benzenamine, 2,4,6-trichloro, aniline, 2,4,6-trichloro, sym-trichloroaniline, 2,4,6-trichlorobenzenamine, 2,4,6-trichlorophenylamine, 1-amino-2,4,6-trichlorobenzene, unii-j7ic72n9b0, ccris 601, pubchem9974
PubChem CID 12471
IUPAC Name 2,4,6-trichloroaniline
SMILES C1=C(C=C(C(=C1Cl)N)Cl)Cl

Specifications

Melting Point 78°C
Color Red-White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
UN Number 3077
Formula Weight 196.46
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,4,6-Trichloroaniline
EINECSNumber (3)-0290
RTECSNumber BZ0250000
TSCA Yes