missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichloroaniline, 98+%

CAS: 634-93-5 | C6H4Cl3N | 196.455 g/mol

Supplier:  Thermo Scientific Chemicals A1904422

 View more versions of this product

Catalog No. AAA1904422


Only null left
Explore available promotions

Description

Description

Tetrachloronitrobenzene was prepared from 2:4:6-trichloroaniline,which was converted by a Sandmeyer reaction into 1:2:3:5- tetrachlorobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrachloronitrobenzene was prepared from 2:4:6-trichloroaniline,which was converted by a Sandmeyer reaction into 1:2:3:5- tetrachlorobenzene.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

2,4,6-Trichloroaniline
634-93-5
100 g
MFCD00007663
879568
Insoluble in water.
C1=C(C=C(C(=C1Cl)N)Cl)Cl
196.455
196.46
75°C to 79°C
262°C
C6H4Cl3N
UN2018
s-trichloroaniline, benzenamine, 2,4,6-trichloro, aniline, 2,4,6-trichloro, sym-trichloroaniline, 2,4,6-trichlorobenzenamine, 2,4,6-trichlorophenylamine, 1-amino-2,4,6-trichlorobenzene, unii-j7ic72n9b0, ccris 601, pubchem9974
NATVSFWWYVJTAZ-UHFFFAOYSA-N
2,4,6-trichloroaniline
12471
≥98%

Chemical Identifiers

634-93-5
196.455
NATVSFWWYVJTAZ-UHFFFAOYSA-N
12471
C1=C(C=C(C(=C1Cl)N)Cl)Cl
C6H4Cl3N
MFCD00007663
s-trichloroaniline, benzenamine, 2,4,6-trichloro, aniline, 2,4,6-trichloro, sym-trichloroaniline, 2,4,6-trichlorobenzenamine, 2,4,6-trichlorophenylamine, 1-amino-2,4,6-trichlorobenzene, unii-j7ic72n9b0, ccris 601, pubchem9974
2,4,6-trichloroaniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c

H301+H311+H331-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROANILINES, SOLID

EINECSNumber : 211-219-8

RTECSNumber : BZ0250000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only