Learn More
2,4,6-Tribromo-3-hydroxybenzaldehyde, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0865609
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2,4,6-Tribromo-3-hydroxybenzaldehyde | |
| 119°C to 122°C | |
| MFCD00017320 | |
| 2098037 | |
| 3-hydroxy-2,4,6-tribromobenzaldehyde, benzaldehyde, 3-hydroxy-2,4,6-tribromo, benzaldehyde,2,4,6-tribromo-3-hydroxy, acmc-1cj0g, 3-08-00-00209 beilstein handbook reference, 2,4,6-tris bromanyl-3-oxidanyl-benzaldehyde | |
| OC1=C(Br)C=C(Br)C(C=O)=C1Br | |
| 358.81 | |
| 358.81 |
| 2737-22-6 | |
| C7H3Br3O2 | |
| 10 g | |
| Air Sensitive | |
| FAWOIFAFFUDNJX-UHFFFAOYSA-N | |
| 2,4,6-tribromo-3-hydroxybenzaldehyde | |
| 200803 | |
| 98% |
Chemical Identifiers
| 2737-22-6 | |
| 358.81 | |
| FAWOIFAFFUDNJX-UHFFFAOYSA-N | |
| 200803 | |
| OC1=C(Br)C=C(Br)C(C=O)=C1Br |
| C7H3Br3O2 | |
| MFCD00017320 | |
| 3-hydroxy-2,4,6-tribromobenzaldehyde, benzaldehyde, 3-hydroxy-2,4,6-tribromo, benzaldehyde,2,4,6-tribromo-3-hydroxy, acmc-1cj0g, 3-08-00-00209 beilstein handbook reference, 2,4,6-tris bromanyl-3-oxidanyl-benzaldehyde | |
| 2,4,6-tribromo-3-hydroxybenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : CU6900000
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only