missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triamino-5-nitrosopyrimidine 98.0+%, TCI America™

Catalog No. p-7136921
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17055G 5 g
T170525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17055G Supplier TCI America Supplier No. T17055G
Only null left

Chemical Identifiers

CAS 1006-23-1
Molecular Formula C4H6N6O
Molecular Weight (g/mol) 154.13
MDL Number MFCD00006096
InChI Key XLQQJSWJHHKLOK-UHFFFAOYSA-N
PubChem CID 70504
IUPAC Name 5-nitrosopyrimidine-2,4,6-triamine
SMILES NC1=NC(N)=C(N=O)C(N)=N1

Specifications

Color Red
Quantity 5 g
Formula Weight 154.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Triamino-5-nitrosopyrimidine