missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tri(4-pyridyl)-1,3,5-triazine (purified by sublimation) 97.0+%, TCI America™

Catalog No. p-7137029
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19371G 1 g
T19375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19371G Supplier TCI America Supplier No. T19371G
Only null left

Chemical Identifiers

CAS 42333-78-8
Molecular Formula C18H12N6
Molecular Weight (g/mol) 312.336
MDL Number MFCD00129048
InChI Key CBMYFVSIIYILRH-UHFFFAOYSA-N
PubChem CID 317666
IUPAC Name 2,4,6-tripyridin-4-yl-1,3,5-triazine
SMILES C1=CN=CC=C1C2=NC(=NC(=N2)C3=CC=NC=C3)C4=CC=NC=C4

Specifications

Quantity 1 g
Formula Weight 312.34
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tri(4-pyridyl)-1,3,5-triazine (purified by sublimation)