missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Tri(4-pyridyl)-1,3,5-triazine (purified by sublimation) 97.0+%, TCI America™
Specifications
Specifications
| Quantity | 5 g |
| Formula Weight | 312.34 |
| Percent Purity | ≥97.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,4,6-Tri(4-pyridyl)-1,3,5-triazine (purified by sublimation) |
Safety and Handling
| TSCA | No |