missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Tri(2-pyridyl)-1,3,5-triazine 98.0+%, TCI America™
Supplier: TCI America T053025G
Specifications
| 2,4,6-Tri(2-pyridyl)-1,3,5-triazine | |
| White-Yellow | |
| MFCD00006045 | |
| 2,4,6-tripyridyl-s-triazine, 2,4,6-tris 2-pyridyl-s-triazine, 2,4,6-tri 2-pyridyl-1,3,5-triazine, terpyridyl-s-triazine, 2,4,6-tri 2-pyridyl-s-triazine, tptz iron reagent, 1,3,5-triazine, 2,4,6-tri-2-pyridinyl, tri-2-pyridyl-s-triazine, s-triazine, tri-2-pyridyl, 2,4,6-tris 2-pyridyl-1,3,5-triazine | |
| C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1 | |
| 312.34 | |
| 312.34 | |
| Crystalline Powder |
| 3682-35-7 | |
| C18H12N6 | |
| 25 g | |
| KMVWNDHKTPHDMT-UHFFFAOYSA-N | |
| tris(pyridin-2-yl)-1,3,5-triazine | |
| 77258 | |
| ≥98.0% (T) |
Chemical Identifiers
| 3682-35-7 | |
| 312.34 | |
| KMVWNDHKTPHDMT-UHFFFAOYSA-N | |
| 77258 | |
| C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1 |
| C18H12N6 | |
| MFCD00006045 | |
| 2,4,6-tripyridyl-s-triazine, 2,4,6-tris 2-pyridyl-s-triazine, 2,4,6-tri 2-pyridyl-1,3,5-triazine, terpyridyl-s-triazine, 2,4,6-tri 2-pyridyl-s-triazine, tptz iron reagent, 1,3,5-triazine, 2,4,6-tri-2-pyridinyl, tri-2-pyridyl-s-triazine, s-triazine, tri-2-pyridyl, 2,4,6-tris 2-pyridyl-1,3,5-triazine | |
| tris(pyridin-2-yl)-1,3,5-triazine |
Safety and Handling
RTECSNumber : XZ2050000
TSCA : Yes