missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tri(2-pyridyl)-1,3,5-triazine 98.0+%, TCI America™

Catalog No. T05301G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T05301G 1 g
T05305G 5 g
T053025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T05301G Supplier TCI America Supplier No. T05301G

Chemical Identifiers

CAS 3682-35-7
Molecular Formula C18H12N6
Molecular Weight (g/mol) 312.34
MDL Number MFCD00006045
InChI Key KMVWNDHKTPHDMT-UHFFFAOYSA-N
Synonym 2,4,6-tripyridyl-s-triazine, 2,4,6-tris 2-pyridyl-s-triazine, 2,4,6-tri 2-pyridyl-1,3,5-triazine, terpyridyl-s-triazine, 2,4,6-tri 2-pyridyl-s-triazine, tptz iron reagent, 1,3,5-triazine, 2,4,6-tri-2-pyridinyl, tri-2-pyridyl-s-triazine, s-triazine, tri-2-pyridyl, 2,4,6-tris 2-pyridyl-1,3,5-triazine
PubChem CID 77258
IUPAC Name tris(pyridin-2-yl)-1,3,5-triazine
SMILES C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 312.34
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tri(2-pyridyl)-1,3,5-triazine
RTECSNumber XZ2050000
TSCA Yes