Learn More
2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 98%
CAS: 3682-35-7 | C18H12N6 | 312.34 g/mol
Supplier: Thermo Scientific Chemicals A1720122
Description
Reagent for the colorimetric determination of iron2,4,6-Tri(2-pyridyl)-1,3,5-triazine is used in the spectrophotometric determination of iron. It acts as a tridendate ligand and form coordination complexes with lanthanides and rhodium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReagent for the colorimetric determination of iron2,4,6-Tri(2-pyridyl)-1,3,5-triazine is used in the spectrophotometric determination of iron. It acts as a tridendate ligand and form coordination complexes with lanthanides and rhodium.
Solubility
Soluble in methanol.
Notes
Incompatible with strong oxidizing agents and strong acids. Store in a cool place.
Specifications
| 2,4,6-Tri(2-pyridyl)-1,3,5-triazine | |
| 248°C to 252°C | |
| MFCD00006045 | |
| 282581 | |
| 2,4,6-tripyridyl-s-triazine, 2,4,6-tris 2-pyridyl-s-triazine, 2,4,6-tri 2-pyridyl-1,3,5-triazine, terpyridyl-s-triazine, 2,4,6-tri 2-pyridyl-s-triazine, tptz iron reagent, 1,3,5-triazine, 2,4,6-tri-2-pyridinyl, tri-2-pyridyl-s-triazine, s-triazine, tri-2-pyridyl, 2,4,6-tris 2-pyridyl-1,3,5-triazine | |
| KMVWNDHKTPHDMT-UHFFFAOYSA-N | |
| tris(pyridin-2-yl)-1,3,5-triazine | |
| 77258 | |
| 98% |
| 3682-35-7 | |
| C18H12N6 | |
| 100 g | |
| 14,9750 | |
| Soluble in methanol. | |
| C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1 | |
| 312.34 | |
| 312.34 |
Chemical Identifiers
| 3682-35-7 | |
| 312.34 | |
| KMVWNDHKTPHDMT-UHFFFAOYSA-N | |
| 77258 |
| C18H12N6 | |
| MFCD00006045 | |
| 2,4,6-tripyridyl-s-triazine, 2,4,6-tris 2-pyridyl-s-triazine, 2,4,6-tri 2-pyridyl-1,3,5-triazine, terpyridyl-s-triazine, 2,4,6-tri 2-pyridyl-s-triazine, tptz iron reagent, 1,3,5-triazine, 2,4,6-tri-2-pyridinyl, tri-2-pyridyl-s-triazine, s-triazine, tri-2-pyridyl, 2,4,6-tris 2-pyridyl-1,3,5-triazine | |
| C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 222-965-9
RTECSNumber : XZ2050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only