missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 98%

Catalog No. AAA1720106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1720106 5 g
AAA1720114 25 g
AAA1720122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1720106 Supplier Thermo Scientific Chemicals Supplier No. A1720106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3682-35-7 | C18H12N6 | 312.34 g/mol

Reagent for the colorimetric determination of iron2,4,6-Tri(2-pyridyl)-1,3,5-triazine is used in the spectrophotometric determination of iron. It acts as a tridendate ligand and form coordination complexes with lanthanides and rhodium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for the colorimetric determination of iron2,4,6-Tri(2-pyridyl)-1,3,5-triazine is used in the spectrophotometric determination of iron. It acts as a tridendate ligand and form coordination complexes with lanthanides and rhodium.

Solubility
Soluble in methanol.

Notes
Incompatible with strong oxidizing agents and strong acids. Store in a cool place.

Chemical Identifiers

CAS 3682-35-7
Molecular Formula C18H12N6
Molecular Weight (g/mol) 312.34
MDL Number MFCD00006045
InChI Key KMVWNDHKTPHDMT-UHFFFAOYSA-N
Synonym 2,4,6-tripyridyl-s-triazine, 2,4,6-tris 2-pyridyl-s-triazine, 2,4,6-tri 2-pyridyl-1,3,5-triazine, terpyridyl-s-triazine, 2,4,6-tri 2-pyridyl-s-triazine, tptz iron reagent, 1,3,5-triazine, 2,4,6-tri-2-pyridinyl, tri-2-pyridyl-s-triazine, s-triazine, tri-2-pyridyl, 2,4,6-tris 2-pyridyl-1,3,5-triazine
PubChem CID 77258
SMILES C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1

Specifications

Melting Point 248°C to 252°C
Quantity 5 g
Beilstein 282581
Merck Index 14,9750
Solubility Information Soluble in methanol.
IUPAC Name 2,4,6-tripyridin-2-yl-1,3,5-triazine
Formula Weight 312.34
Percent Purity 98%
Chemical Name or Material 2,4,6-Tri(2-pyridyl)-1,3,5-triazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-965-9
RTECSNumber XZ2050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only