missing translation for 'onlineSavingsMsg'
Learn More

2,4,6,8-Tetramethylcyclotetrasiloxane 95.0+%, TCI America™

Catalog No. p-7137090
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Catalog No. Quantity
T20765G 5 g
T207625G 25 g
T2076100G 100 g
3 options

Catalog No. T20765G

Supplier: TCI America T20765G

Only null left

Chemical Identifiers

CAS 2370-88-9
Molecular Formula C4H12O4Si4
Molecular Weight (g/mol) 236.476
MDL Number MFCD00039567
InChI Key WZJUBBHODHNQPW-UHFFFAOYSA-N
Synonym 2,4,6,8-tetramethylcyclotetrasiloxane, 1,3,5,7-tetramethylcyclotetrasiloxane, tetramethylcyclotetrasiloxane, tmcts, cyclotetrasiloxane, 2,4,6,8-tetramethyl, 2,4,6,8-tetramethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, 1,2,5,7-tetramethylcyclotetrasiloxane, 2,4,6,8-tetramethyl-cyclotetrasiloxan, 2,4,6,8-tetramethylcyclotetrasiloxne, 1,3,5,7-tetramethyl cyclotetrasiloxane
PubChem CID 6327421
IUPAC Name 2,4,6,8-tetramethyl-5,7-dioxa-1,3-dioxonia-2,4-disila-6,8-disilanidacycloocta-1,3-diene
SMILES C[Si-]1O[Si-]([O+]=[Si]([O+]=[Si](O1)C)C)C

Specifications

Color Colorless
Boiling Point 134°C
Quantity 5 g
UN Number 1993
Formula Weight 240.51
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,6,8-Tetramethylcyclotetrasiloxane
EINECSNumber (7)-0477
TSCA Yes
Recommended Storage Refrigerator