missing translation for 'onlineSavingsMsg'
Learn More

2,4,6,8-Tetramethylcyclotetrasiloxane, 99%

Catalog No. AAL1664222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1664222 100 g
AAL1664214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1664222 Supplier Thermo Scientific Chemicals Supplier No. L1664222

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2370-88-9 | C4H12O4Si4 | 236.476 g/mol

Tetramethyl-cyclotetrasiloxane is a good impregnant of photoelectric material. It is used to manufacture of modified siloxane with defined hydrogen content and chain quantity. It is also used in production of silicone polymers and acts as a precursor for the deposition of polysiloxane, cyclic siloxane, silicon dioxide, oxycarbide thin films with low dielectric constant for microelectronics and semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethyl-cyclotetrasiloxane is a good impregnant of photoelectric material. It is used to manufacture of modified siloxane with defined hydrogen content and chain quantity. It is also used in production of silicone polymers and acts as a precursor for the deposition of polysiloxane, cyclic siloxane, silicon dioxide, oxycarbide thin films with low dielectric constant for microelectronics and semiconductors.

Solubility
Miscible with most organic solvents. Immiscible with water.

Notes
Incompatible with oxidizing agents, bases and acids. Store in a cool place.

Chemical Identifiers

CAS 2370-88-9
Molecular Formula C4H12O4Si4
Molecular Weight (g/mol) 236.476
MDL Number MFCD00039567
InChI Key WZJUBBHODHNQPW-UHFFFAOYSA-N
Synonym 2,4,6,8-tetramethylcyclotetrasiloxane, 1,3,5,7-tetramethylcyclotetrasiloxane, tetramethylcyclotetrasiloxane, tmcts, cyclotetrasiloxane, 2,4,6,8-tetramethyl, 2,4,6,8-tetramethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, 1,2,5,7-tetramethylcyclotetrasiloxane, 2,4,6,8-tetramethyl-cyclotetrasiloxan, 2,4,6,8-tetramethylcyclotetrasiloxne, 1,3,5,7-tetramethyl cyclotetrasiloxane
PubChem CID 6327421
IUPAC Name 2,4,6,8-tetramethyl-5,7-dioxa-1,3-dioxonia-2,4-disila-6,8-disilanidacycloocta-1,3-diene
SMILES C[Si-]1O[Si-]([O+]=[Si]([O+]=[Si](O1)C)C)C

Specifications

Boiling Point 134°C to 135°C
Melting Point -69°C
Quantity 100 g
Flash Point 25°C (77°F)
Refractive Index 1.387
Sensitivity Moisture sensitive
Density 0.991
UN Number UN1993
Beilstein 1787074
Solubility Information Miscible with most organic solvents. Immiscible with water.
Formula Weight 240.51
Percent Purity 99%
Chemical Name or Material 2,4,6,8-Tetramethylcyclotetrasiloxane
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H319
Flammable liquid and vapor.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 219-137-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only