missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trimethylthiazole 97.0+%, TCI America™

Catalog No. T10685G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10685G 5 g
T106825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10685G Supplier TCI America Supplier No. T10685G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13623-11-5
Molecular Formula C6H9NS
Molecular Weight (g/mol) 127.205
MDL Number MFCD00005332
InChI Key BAMPVSWRQZNDQC-UHFFFAOYSA-N
Synonym 2,4,5-trimethylthiazole, trimethylthiazole, trimethyl thiazole, thiazole, trimethyl, thiazole, 2,4,5-trimethyl, trimethyl-1,3-thiazole, 2,4,5-trimethyl thiazole, thiazole, 2,4,5-trimethyl-8ci 9ci, fema no. 3325, trimethyl-thiazole
PubChem CID 61653
ChEBI CHEBI:78738
IUPAC Name 2,4,5-trimethyl-1,3-thiazole
SMILES CC1=C(SC(=N1)C)C

Specifications

Color Yellow
Boiling Point 167°C
Quantity 5 g
UN Number 1993
Formula Weight 127.21
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,4,5-Trimethylthiazole