missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,5-Trimethyloxazole 98.0+%, TCI America™
$65.24 - $65.24
Chemical Identifiers
| CAS | 20662-84-4 |
|---|---|
| Molecular Formula | C6H9NO |
| Molecular Weight (g/mol) | 111.144 |
| MDL Number | MFCD00005308 |
| InChI Key | ZRLDBDZSLLGDOX-UHFFFAOYSA-N |
| Synonym | 2,4,5-trimethyloxazole, trimethyloxazole, oxazole, trimethyl, unii-b04pf51wxi, 2,4,5-trimethyl oxazole, trimethyl-1,3-oxazole, b04pf51wxi, oxazole, 2,4,5-trimethyl, trimethyl-oxazole, pubchem8636 |
| PubChem CID | 30215 |
| IUPAC Name | 2,4,5-trimethyl-1,3-oxazole |
| SMILES | CC1=C(OC(=N1)C)C |
Chemical Identifiers
| 20662-84-4 | |
| 111.144 | |
| ZRLDBDZSLLGDOX-UHFFFAOYSA-N | |
| 30215 | |
| CC1=C(OC(=N1)C)C |
| C6H9NO | |
| MFCD00005308 | |
| 2,4,5-trimethyloxazole, trimethyloxazole, oxazole, trimethyl, unii-b04pf51wxi, 2,4,5-trimethyl oxazole, trimethyl-1,3-oxazole, b04pf51wxi, oxazole, 2,4,5-trimethyl, trimethyl-oxazole, pubchem8636 | |
| 2,4,5-trimethyl-1,3-oxazole |
Specifications
| 20662-84-4 | |
| 134°C | |
| MFCD00005308 | |
| 1993 | |
| ZRLDBDZSLLGDOX-UHFFFAOYSA-N | |
| 2,4,5-trimethyl-1,3-oxazole | |
| 30215 | |
| ≥98.0% (GC) | |
| 2,4,5-Trimethyloxazole |
| Yellow | |
| C6H9NO | |
| 5 g | |
| 2,4,5-trimethyloxazole, trimethyloxazole, oxazole, trimethyl, unii-b04pf51wxi, 2,4,5-trimethyl oxazole, trimethyl-1,3-oxazole, b04pf51wxi, oxazole, 2,4,5-trimethyl, trimethyl-oxazole, pubchem8636 | |
| CC1=C(OC(=N1)C)C | |
| 111.144 | |
| 111.14 | |
| Liquid |
Safety and Handling
EINECSNumber : (5)-3394
TSCA : No