missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trimethyloxazole, 97%

Catalog No. p-7080690
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1032806 5 g
AAA1032814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1032806 Supplier Thermo Scientific Chemicals Supplier No. A1032806
Only null left

CAS: 20662-84-4 | C6H9NO | 111.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20662-84-4
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.144
MDL Number MFCD00005308
InChI Key ZRLDBDZSLLGDOX-UHFFFAOYSA-N
Synonym 2,4,5-trimethyloxazole, trimethyloxazole, oxazole, trimethyl, unii-b04pf51wxi, 2,4,5-trimethyl oxazole, trimethyl-1,3-oxazole, b04pf51wxi, oxazole, 2,4,5-trimethyl, trimethyl-oxazole, pubchem8636
PubChem CID 30215
IUPAC Name 2,4,5-trimethyl-1,3-oxazole
SMILES CC1=C(OC(=N1)C)C

Specifications

Density 0.964
Boiling Point 133°C to 134°C
Flash Point 33°C (91°F)
Odor Pungent
Refractive Index 1.442
Quantity 5 g
UN Number UN1993
Beilstein 107043
Formula Weight 111.14
Percent Purity 97%
Chemical Name or Material 2,4,5-Trimethyloxazole
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H319-H335
Flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 243-952-4
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only