missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,5-Trifluorophenylboronic acid, 97%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 96% min. (HPLC) |
| Packaging | Glass bottle |
| Linear Formula | F3C6H2B(OH)2 |
| Quantity | 5g |
| Solubility Information | Solubility in water: .. Other solubilities: soluble in methanol |
| Formula Weight | 175.9 |
| Percent Purity | 97% |
| Chemical Name or Material | 2, 4, 5-Trifluorophenylboronic acid |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation. |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash |
RUO – Research Use Only