missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,5-Trifluorobenzenesulfonyl Chloride 98.0+%, TCI America™
$113.71 - $451.10
Chemical Identifiers
| CAS | 220227-21-4 |
|---|---|
| Molecular Formula | C6H2ClF3O2S |
| Molecular Weight (g/mol) | 230.585 |
| MDL Number | MFCD00077600 |
| InChI Key | STCZWXXRRTWRSK-UHFFFAOYSA-N |
| Synonym | 2,4,5-trifluorobenzene-1-sulfonyl chloride, 2,4,5-trifluorobenzenesulphonyl chloride, 2,4,5-trifluorophenyl sulfonyl chloride, 2,4,5-trifluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 2,4,5-trifluoro, pubchem5179, acmc-209fqs, ksc201o4t, chloro 2,4,5-trifluorophenyl sulfone, 2,4,5-trifluorophenylsulfonyl chloride |
| PubChem CID | 2776979 |
| IUPAC Name | 2,4,5-trifluorobenzenesulfonyl chloride |
| SMILES | C1=C(C(=CC(=C1F)S(=O)(=O)Cl)F)F |
Chemical Identifiers
| 220227-21-4 | |
| 230.585 | |
| STCZWXXRRTWRSK-UHFFFAOYSA-N | |
| 2776979 | |
| C1=C(C(=CC(=C1F)S(=O)(=O)Cl)F)F |
| C6H2ClF3O2S | |
| MFCD00077600 | |
| 2,4,5-trifluorobenzene-1-sulfonyl chloride, 2,4,5-trifluorobenzenesulphonyl chloride, 2,4,5-trifluorophenyl sulfonyl chloride, 2,4,5-trifluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 2,4,5-trifluoro, pubchem5179, acmc-209fqs, ksc201o4t, chloro 2,4,5-trifluorophenyl sulfone, 2,4,5-trifluorophenylsulfonyl chloride | |
| 2,4,5-trifluorobenzenesulfonyl chloride |
Specifications
| 220227-21-4 | |
| C6H2ClF3O2S | |
| 1 g | |
| 2,4,5-trifluorobenzene-1-sulfonyl chloride, 2,4,5-trifluorobenzenesulphonyl chloride, 2,4,5-trifluorophenyl sulfonyl chloride, 2,4,5-trifluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 2,4,5-trifluoro, pubchem5179, acmc-209fqs, ksc201o4t, chloro 2,4,5-trifluorophenyl sulfone, 2,4,5-trifluorophenylsulfonyl chloride | |
| C1=C(C(=CC(=C1F)S(=O)(=O)Cl)F)F | |
| 230.585 | |
| 230.59 | |
| Liquid |
| Yellow | |
| MFCD00077600 | |
| 3265 | |
| STCZWXXRRTWRSK-UHFFFAOYSA-N | |
| 2,4,5-trifluorobenzenesulfonyl chloride | |
| 2776979 | |
| ≥98.0% (GC,T) | |
| 2,4,5-Trifluorobenzenesulfonyl Chloride |
Safety and Handling
TSCA : No