missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,5-Trifluorobenzenesulfonyl Chloride 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| UN Number | 3265 |
| Formula Weight | 230.59 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,4,5-Trifluorobenzenesulfonyl Chloride |
Safety and Handling
| TSCA | No |