missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trichlorophenol Sodium Salt 98.0+%, TCI America™

Catalog No. T039125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T039125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T039125G Supplier TCI America Supplier No. T039125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 136-32-3
Molecular Formula C6H2Cl3NaO
Molecular Weight (g/mol) 219.421
MDL Number MFCD00058974
InChI Key KWFOMJTYKROHLX-UHFFFAOYSA-M
PubChem CID 23662264
IUPAC Name sodium;2,4,5-trichlorophenolate
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)[O-].[Na+]

Specifications

Quantity 25 g
Physical Form Crystal-Powder at 20°C
UN Number 2811
Formula Weight 219.42
Percent Purity ≥97.0% (T)
Chemical Name or Material 2,4,5-Trichlorophenol Sodium Salt
EINECSNumber (3)-0931
RTECSNumber SN2750000
TSCA Yes