missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Triamino-6-hydroxypyrimidine Sulfate Hydrate 90.0+%, TCI America™

Catalog No. T033525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T033525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T033525G Supplier TCI America Supplier No. T033525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35011-47-3
Molecular Formula C4H9N5O5S
Molecular Weight (g/mol) 239.206
MDL Number MFCD00012785
InChI Key RSKNEEODWFLVFF-UHFFFAOYSA-N
Synonym 2,4,5-triamino-6-hydroxypyrimidine sulfate, 6-hydroxy-2,4,5-triaminopyrimidine sulfate, 2,5,6-triaminopyrimidin-4 3h-one sulfate, unii-9sz7zx1h0l, sulfuric acid; triaminopyrimidin-4-ol, 2,5,6-triamino-4-pyrimidinol sulfate, 4 1h-pyrimidinone, 2,5,6-triamino-, sulfate, 2,5,6-triaminopyrimidin-4 1h-one sulphate, 2,5,6-triaminopyrimidin-4-ol sulfate, 9sz7zx1h0l
PubChem CID 296856
IUPAC Name sulfuric acid;2,5,6-triamino-1H-pyrimidin-4-one
SMILES C1(=C(NC(=NC1=O)N)N)N.OS(=O)(=O)O

Specifications

Quantity 25 g
Formula Weight 239.21
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Triamino-6-hydroxypyrimidine Sulfate Hydrate