missing translation for 'onlineSavingsMsg'
Learn More

2,4,5,6-Tetrafluoropyrimidine, 95%

Catalog No. AAH32453MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32453MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH32453MD Supplier Thermo Scientific Chemicals Supplier No. H32453MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 767-79-3 | C4F4N2 | 152.052 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 767-79-3
Molecular Formula C4F4N2
Molecular Weight (g/mol) 152.052
MDL Number MFCD10687402
InChI Key KZMWBUVUQLGBBP-UHFFFAOYSA-N
Synonym tetrafluoropyrimidine, pyrimidine, tetrafluoro, pyrimidine,tetrafluoro, perfluoropyrimidine
PubChem CID 136605
IUPAC Name 2,4,5,6-tetrafluoropyrimidine
SMILES C1(=C(N=C(N=C1F)F)F)F

Specifications

Boiling Point 83°C
Quantity 250 mg
UN Number UN3265
Formula Weight 152.05
Percent Purity 95%
Chemical Name or Material 2,4,5,6-Tetrafluoropyrimidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only