missing translation for 'onlineSavingsMsg'
Learn More

2,4,5,6-Tetrachloropyrimidine 98.0+%, TCI America™

Catalog No. t12175g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T12175G Supplier TCI America Supplier No. T12175G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1780-40-1
Molecular Formula C4Cl4N2
Molecular Weight (g/mol) 217.858
MDL Number MFCD00006062
InChI Key GVBHCMNXRKOJRH-UHFFFAOYSA-N
Synonym perchloropyrimidine, pyrimidine, tetrachloro, tetrachloropyrimidine, pyrimidine, 2,4,5,6-tetrachloro, 2,4,5,6-tetrachloro-1,3-pyrimidine, tetrachlorpyrimidin, tetrachloro-pyrimidin, acmc-1bsuq, 2,5,6-tetrachloropyrimidine, 5-23-05-00347 beilstein handbook reference
PubChem CID 15690
IUPAC Name 2,4,5,6-tetrachloropyrimidine
SMILES C1(=C(N=C(N=C1Cl)Cl)Cl)Cl

Specifications

Melting Point 68°C
Quantity 5 g
Formula Weight 217.86
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5,6-Tetrachloropyrimidine
RTECSNumber UV9745000
TSCA Yes