missing translation for 'onlineSavingsMsg'
Learn More

2,4,4-Trimethyl-2-pentene, 97%

Catalog No. AAB2077314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2077314 25 g
AAB2077322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2077314 Supplier Thermo Scientific Chemicals Supplier No. B2077314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107-40-4 | C8H16 | 112.216 g/mol

2,4,4-Trimethyl-2-pentene was used to study the kinetics of liquid-phase hydrogenation of isooctenes on Pd/γ-Al2O3 catalyst in semibatch reactor. It was used in the analysis of butene oligomerate produced over solid and liquid phosphoric acid by GC-MS techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,4-Trimethyl-2-pentene was used to study the kinetics of liquid-phase hydrogenation of isooctenes on Pd/γ-Al2O3 catalyst in semibatch reactor. It was used in the analysis of butene oligomerate produced over solid and liquid phosphoric acid by GC-MS techniques.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 107-40-4
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00008902
InChI Key LAAVYEUJEMRIGF-UHFFFAOYSA-N
Synonym 2,4,4-trimethyl-2-pentene, 2-pentene, 2,4,4-trimethyl, 2,2,4-trimethyl-3-pentene, 2,4,4-trimethylpentene-2, unii-548r0du21i, beta-diisobutylene, propene, 1-tert-butyl-2-methyl, acmc-1cujo, dsstox_cid_6766, dsstox_rid_78208
PubChem CID 7869
IUPAC Name 2,4,4-trimethylpent-2-ene
SMILES CC(=CC(C)(C)C)C

Specifications

Melting Point -106°C
Density 0.72
Boiling Point 103°C to 105°C
Flash Point −1°C (29°F)
Refractive Index 1.416
Quantity 25 g
UN Number UN3295
Solubility Information Insoluble in water.
Formula Weight 112.22
Percent Purity 97%
Chemical Name or Material 2,4,4-Trimethyl-2-pentene
Show More Show Less
Hazard Category H225-H304-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 203-488-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only