missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,4-Trimethyl-2-pentanol 95.0+%, TCI America™
$188.27 - $555.49
Chemical Identifiers
| CAS | 690-37-9 |
|---|---|
| Molecular Formula | C8H18O |
| Molecular Weight (g/mol) | 130.231 |
| MDL Number | MFCD00101611 |
| InChI Key | BSYJHYLAMMJNRC-UHFFFAOYSA-N |
| Synonym | 2-Hydroxy-2,4,4-trimethylpentane |
| PubChem CID | 94740 |
| IUPAC Name | 2,4,4-trimethylpentan-2-ol |
| SMILES | CC(C)(C)CC(C)(C)O |
Chemical Identifiers
| 690-37-9 | |
| 130.231 | |
| BSYJHYLAMMJNRC-UHFFFAOYSA-N | |
| 94740 | |
| CC(C)(C)CC(C)(C)O |
| C8H18O | |
| MFCD00101611 | |
| 2-Hydroxy-2,4,4-trimethylpentane | |
| 2,4,4-trimethylpentan-2-ol |
Specifications
| 690-37-9 | |
| C8H18O | |
| 1 g | |
| 2-Hydroxy-2,4,4-trimethylpentane | |
| CC(C)(C)CC(C)(C)O | |
| 130.231 | |
| 130.23 | |
| Liquid |
| Yellow | |
| MFCD00101611 | |
| 1987 | |
| BSYJHYLAMMJNRC-UHFFFAOYSA-N | |
| 2,4,4-trimethylpentan-2-ol | |
| 94740 | |
| ≥95.0% (GC) | |
| 2,4,4-Trimethyl-2-pentanol |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : No