missing translation for 'onlineSavingsMsg'
Learn More

2,4,4-Trimethyl-1-pentene 98.0+%, TCI America™

Catalog No. p-7136383
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T066525ML 25 mL
T0665100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T066525ML Supplier TCI America Supplier No. T066525ML
Only null left

Chemical Identifiers

CAS 107-39-1
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00008855
InChI Key FXNDIJDIPNCZQJ-UHFFFAOYSA-N
Synonym 2,4,4-trimethyl-1-pentene, diisobutylene, pentene, 2,4,4-trimethyl, 2,4,4-trimethylpentene, 1-pentene, 2,4,4-trimethyl, 2,2,4-trimethyl-4-pentene, unii-n69l73advf, 1-methyl-1-neopentylethylene, ccris 9103, 2,4,4-trimethylpentene-1
PubChem CID 7868
IUPAC Name 2,4,4-trimethylpent-1-ene
SMILES CC(=C)CC(C)(C)C

Specifications

Melting Point -93°C
Color Colorless
Boiling Point 100°C
Quantity 25 mL
UN Number 2050
Formula Weight 112.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,4-Trimethyl-1-pentene
EINECSNumber (2)-0024
RTECSNumber SB2715000
TSCA Yes