Learn More
2,4,4-Trimethyl-1-pentene, 99%
CAS: 107-39-1 | C8H16 | 112.21 g/mol
$238.13 - $238.13
Chemical Identifiers
| CAS | 107-39-1 |
|---|---|
| Molecular Formula | C8H16 |
| Molecular Weight (g/mol) | 112.21 |
| MDL Number | MFCD00008855 |
| InChI Key | FXNDIJDIPNCZQJ-UHFFFAOYSA-N |
| Synonym | 2,4,4-trimethyl-1-pentene, diisobutylene, pentene, 2,4,4-trimethyl, 2,4,4-trimethylpentene, 1-pentene, 2,4,4-trimethyl, 2,2,4-trimethyl-4-pentene, unii-n69l73advf, 1-methyl-1-neopentylethylene, ccris 9103, 2,4,4-trimethylpentene-1 |
| PubChem CID | 7868 |
| IUPAC Name | 2,4,4-trimethylpent-1-ene |
| SMILES | CC(=C)CC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC140181000
|
Thermo Scientific Chemicals
140181000 |
100 mL | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 107-39-1 | |
| 112.21 | |
| FXNDIJDIPNCZQJ-UHFFFAOYSA-N | |
| 7868 | |
| CC(=C)CC(C)(C)C |
| C8H16 | |
| MFCD00008855 | |
| 2,4,4-trimethyl-1-pentene, diisobutylene, pentene, 2,4,4-trimethyl, 2,4,4-trimethylpentene, 1-pentene, 2,4,4-trimethyl, 2,2,4-trimethyl-4-pentene, unii-n69l73advf, 1-methyl-1-neopentylethylene, ccris 9103, 2,4,4-trimethylpentene-1 | |
| 2,4,4-trimethylpent-1-ene |
Specifications
| 107-39-1 | |
| 100.0 | |
| Colorless | |
| 101°C to 102°C | |
| Authentic | |
| Glass bottle | |
| 1.4070 to 1.409 | |
| MFCD00008855 | |
| 0.7 | |
| Solubility in water: immiscible with water | |
| CC(=C)CC(C)(C)C | |
| 112.21 | |
| 112.21 | |
| Liquid |
| 98.5 | |
| -101°C | |
| 0.7000g/mL | |
| -6°C | |
| 99% | |
| C8H16 | |
| (CH3)3CCH2C(CH3)=CH2 | |
| 100 mL | |
| 2,4,4-trimethyl-1-pentene, diisobutylene, pentene, 2,4,4-trimethyl, 2,4,4-trimethylpentene, 1-pentene, 2,4,4-trimethyl, 2,2,4-trimethyl-4-pentene, unii-n69l73advf, 1-methyl-1-neopentylethylene, ccris 9103, 2,4,4-trimethylpentene-1 | |
| FXNDIJDIPNCZQJ-UHFFFAOYSA-N | |
| 2,4,4-trimethylpent-1-ene | |
| 7868 | |
| 99% | |
| 2, 4, 4-Trimethyl-1-pentene |
Safety and Handling
GHS H Statement:
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 203-486-4
RUO – Research Use Only