missing translation for 'onlineSavingsMsg'
Learn More

2,4,4-Trimethyl-1-pentene, 99%

Catalog No. AAB2018722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2018722 100 g
AAB2018714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2018722 Supplier Thermo Scientific Chemicals Supplier No. B2018722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 107-39-1 | C8H16 | 112.216 g/mol

2,4,4-Trimethyl-1-pentene is used in the manufacturing of chemicals, adhesives, sealant chemicals, fuels and fuel additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,4-Trimethyl-1-pentene is used in the manufacturing of chemicals, adhesives, sealant chemicals, fuels and fuel additives.

Solubility
Miscible with acetone. Partially soluble in methanol, diethyl ether, benzene, chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agent.

Chemical Identifiers

CAS 107-39-1
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00008855
InChI Key FXNDIJDIPNCZQJ-UHFFFAOYSA-N
Synonym 2,4,4-trimethyl-1-pentene, diisobutylene, pentene, 2,4,4-trimethyl, 2,4,4-trimethylpentene, 1-pentene, 2,4,4-trimethyl, 2,2,4-trimethyl-4-pentene, unii-n69l73advf, 1-methyl-1-neopentylethylene, ccris 9103, 2,4,4-trimethylpentene-1
PubChem CID 7868
IUPAC Name 2,4,4-trimethylpent-1-ene
SMILES CC(=C)CC(C)(C)C

Specifications

Melting Point -94°C
Density 0.715
Boiling Point 101°C to 102°C
Flash Point −6°C (21°F)
Odor Petrol-like
Refractive Index 1.4085
Quantity 100 g
UN Number UN2050
Beilstein 1098309
Solubility Information Miscible with acetone. Partially soluble in methanol,diethyl ether,benzene,chloroform. Immiscible with water.
Formula Weight 112.22
Percent Purity 99%
Chemical Name or Material 2,4,4-Trimethyl-1-pentene
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIISOBUTYLENE, ISOMERIC COMPOUNDS
EINECSNumber 203-486-4
RTECSNumber SB2717300
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only