missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. S311500
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
10 kg
50 kg
Packaging:
Fiber Drum
Glass Bottle
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S311500 500 g Poly Bottle
S311100 100 g Glass Bottle
S3113 3 kg Poly Bottle
S31110 10 kg Poly Pail
S31150KG 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S311500 Supplier Thermo Fisher Scientific Supplier No. S311 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Decomposition Information >252°C
Melting Point 252°C
Color White
pH 4 to 6
Assay Percent Range 99.0 to 101.0 %
Packaging Poly Bottle
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2Na)2·2H2O
Quantity 500 g
Solubility Information 0.005% max. Insoluble Matter
Formula Weight 372.24
Percent Purity 99.0 to 101.0%
Grade Certified ACS
Physical Form Solid
Chemical Name or Material Ethylenediamine Tetraacetic Acid, Disodium Salt Dihydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:1
Instability:0