missing translation for 'onlineSavingsMsg'
Learn More

2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene 98.0+%, TCI America™

Catalog No. p-7137478
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
T29241G 1 g
T29245G 5 g
2 options

Catalog No. T29241G

Supplier: TCI America T29241G

Only null left

Chemical Identifiers

CAS 193978-23-3
Molecular Formula C10H15BO2S
Molecular Weight (g/mol) 210.098
MDL Number MFCD05663878
InChI Key FFZHICFAHSDFKZ-UHFFFAOYSA-N
Synonym 2-thiopheneboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-thiophen-2-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-thienyl-1,3,2-dioxaborolane, thiophene-2-boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophene, thiophene-2-boronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-thienyl, 4,4,5,5-tetramethyl-2-thien-2-yl-1,3,2-dioxaborolane, pubchem18441
PubChem CID 10703628
IUPAC Name 4,4,5,5-tetramethyl-2-thiophen-2-yl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CS2

Specifications

Melting Point 68°C
Color White-Yellow
Quantity 1 g
Formula Weight 210.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene