missing translation for 'onlineSavingsMsg'
Learn More

2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole 98.0+%, TCI America™

Catalog No. T34415G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34415G 5 g
T34411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34415G Supplier TCI America Supplier No. T34415G
Only null left

Chemical Identifiers

CAS 1242412-60-7
Molecular Formula C18H20BNO2
Molecular Weight (g/mol) 293.173
MDL Number MFCD07784368
InChI Key RLSJGSFDSSYNPL-UHFFFAOYSA-N
Synonym 2-(9H-Carbazol-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, (9H-Carbazol-2-yl)boronic Acid Pinacol Ester
PubChem CID 46738013
IUPAC Name 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4N3

Specifications

Melting Point 197°C
Color White-Yellow
Quantity 5 g
Formula Weight 293.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole