missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]-1,2-oxazinane, ≥97%, Thermo Scientific™

Catalog No. CC45539DA
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC45539DA 1g Amber Glass Bottle
CC45539CB 250mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC45539DA Supplier Thermo Scientific Chemicals Supplier No. CC45539DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 859833-22-0
Molecular Formula C18H28BNO2
Molecular Weight (g/mol) 301.24
MDL Number MFCD08271932
InChI Key HJKVRMHVLINDRH-UHFFFAOYSA-N
Synonym 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperidine, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 4-1-piperidinylmethyl benzeneboronic acid pinacol ester, piperidine,1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl, amtb120, 4-piperidinomethyl phenylboronic acid pinacol ester, 4-piperidinomethyl phenylboronic acid, pinacol ester, 4-piperidin-1-ylmethyl phenyl boronic acid pinacol ester
PubChem CID 18525779
IUPAC Name 1-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperidine
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(CN2CCCCC2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 301.24
Percent Purity 97%
Chemical Name or Material 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperidine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.