missing translation for 'onlineSavingsMsg'
Learn More

2-[4(1H)-Pyridinylidene]indan-1,3-dione, TCI America™

Catalog No. P11245G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P11245G Supplier TCI America Supplier No. P11245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10478-99-6
Molecular Formula C14H9NO2
Molecular Weight (g/mol) 223.23
MDL Number MFCD00142802,MFCD00444488
InChI Key RFJVOUZINWUTIG-UHFFFAOYSA-N
Synonym 2-[4(1H)-Pyridylidene]indan-1,3-dione, gamma-Pyrophthalone
PubChem CID 5389994
IUPAC Name 2-(1,4-dihydropyridin-4-ylidene)-2,3-dihydro-1H-indene-1,3-dione
SMILES O=C1C2=CC=CC=C2C(=O)C1=C1C=CNC=C1

Specifications

Quantity 5 g
Formula Weight 223.23
Physical Form Crystalline Powder
Chemical Name or Material 2-[4(1H)-Pyridinylidene]indan-1,3-dione