missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Thiophenedicarboxylic Anhydride 98.0+%, TCI America™
$251.65 - $758.67
Chemical Identifiers
| CAS | 6007-83-6 |
|---|---|
| Molecular Formula | C6H2O3S |
| Molecular Weight (g/mol) | 154.139 |
| MDL Number | MFCD09832453 |
| InChI Key | FIDKFEIEZJGDBE-UHFFFAOYSA-N |
| PubChem CID | 222951 |
| IUPAC Name | thieno[2,3-c]furan-4,6-dione |
| SMILES | C1=CSC2=C1C(=O)OC2=O |
Chemical Identifiers
| 6007-83-6 | |
| 154.139 | |
| FIDKFEIEZJGDBE-UHFFFAOYSA-N | |
| thieno[2,3-c]furan-4,6-dione |
| C6H2O3S | |
| MFCD09832453 | |
| 222951 | |
| C1=CSC2=C1C(=O)OC2=O |
Specifications
| 6007-83-6 | |
| White-Yellow | |
| MFCD09832453 | |
| FIDKFEIEZJGDBE-UHFFFAOYSA-N | |
| thieno[2,3-c]furan-4,6-dione | |
| 222951 | |
| ≥98.0% (GC,T) | |
| 2,3-Thiophenedicarboxylic Anhydride |
| 142°C | |
| C6H2O3S | |
| 1 g | |
| C1=CSC2=C1C(=O)OC2=O | |
| 154.139 | |
| 154.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No